Learn More
Thermo Scientific™ Spring Loaded Transfer Line Nut for ISQ and TSQ
Product Code. 15392155 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
3-Port SilFlow MCD Device
Swap-IT
Thermo Scientific ISQ and TSQ
Description:
Blanking Ferrule
Chromatography Accessories
Spring Loaded Transfer Nut
Unit Size:
Each
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Description Unit Size
15392155 Thermo Scientific ISQ and TSQ Spring Loaded Transfer Nut Each
15297488 3-Port SilFlow MCD Device Blanking Ferrule Pack of 5
11567761 Swap-IT Chromatography Accessories Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15392155 Supplier Thermo Scientific™ Supplier No. 1R1204340010
Estimated Shipment: 03-12-2026
Make column changeovers quick and easy. Use this GC-MS interface system to switch capillary columns without the need to vent your mass spectrometer (MS).
TRUSTED_SUSTAINABILITY
Specifications
Description Spring Loaded Transfer Nut
For Use With (Equipment) Thermo Scientific ISQ and TSQ
Quantity Each