missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUSD2 Monoclonal Antibody (944827)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561984
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
May be a cytokine receptor for C10ORF99. May be a tumor suppressor; together with C10ORF99 has a growth inhibitory effect on colon cancer cells which includes G1 cell cycle arrest (PubMed:25351403). May play a role in breast tumorigenesis (PubMed:23131994).
Specifications
| SUSD2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9UGT4 | |
| SUSD2 | |
| Human embryonic kidney cell line HEK293-derived human SUSD2, Met1-Ala785. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| 944827 | |
| Unconjugated | |
| SUSD2 | |
| BK65A6.2; SUSD2; Sushi domain (SCR repeat) containing; sushi domain containing 2; sushi domain-containing protein 2; testicular tissue protein Li 190 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 56241 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction