missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUSD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-61129-0.025mg
This item is not returnable.
View return policy
Description
SUSD1 Polyclonal specifically detects SUSD1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| SUSD1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| SUSD1 | |
| SUSD1 antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human SUSD1. Amino Acid Squence: LRITSEWNKVRRHS | |
| Affinity Purified | |
| RUO | |
| 64420 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 | |
| RP11-4O1.1, sushi domain containing 1 | |
| Rabbit | |
| 82 kDa | |
| 0.025 mg | |
| Primary | |
| SUSD1 Antibody is predicted to not cross-react with other SUSDfamiliy members. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction