missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUPV3L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | SUPV3L1 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16870744
|
Proteintech
12826-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16860744
|
Proteintech
12826-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SUV3, also known as SUPV3L1 (suppressor of var1, 3-like 1), is a 786 amino acid protein that localizes to both the nucleus and the mitochondrial matrix and contains one helicase C-terminal domain, as well as one helicase ATP-binding domain. Expressed in a broad range of tissues, SUV3 interacts with HBXIP and functions as an ATPase DNA/RNA helicase that uses magnesium as a cofactor to catalyze the unwinding of DNA/RNA and RNA/RNA duplexes, thereby playing a role in DNA replication and transcriptional initiation. SUV3 exhibits optimal activity at a pH of 5 and, in addition to its helicase activity, is thought to protect cells from apoptosis and participate in maintaining mitochondrial homeostasis.Specifications
| SUPV3L1 | |
| Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q5EBA1, Q80YD1, Q8IYB8 | |
| 294385, 338359, 6832 | |
| SUPV3L1 Fusion Protein Ag3542 | |
| Primary | |
| -20°C | |
| SUPV3L1 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| SUPV3L1, SUV3, SUV3 like protein 1 | |
| Supv3l1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title