Learn More
Labcon™ SuperSlik™ 20 μL Low Retention Pipette Tips for Rainin™ LTS Pipettors
GREENER_CHOICE
Product Code. 16681913
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bagged
Rack, 96
Refill
Unit Size:
Pack of 1000
Pack of 1056
Pack of 960
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Format unitSize
16681913 Non-sterile Refill Pack of 1056
16691913 Non-sterile Rack, 96 Pack of 960
16601923 Sterile Rack, 96 Pack of 960
17507836 Non-sterile Bagged Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16681913 Supplier Labcon™ Supplier No. 10662600009
Edge
These low retention pipette tips with UltraFine™ points are available in resealable bags, racks, or refills.
  • Fits 20 μL Rainin™ LTS style pipettes
  • Low mounting and ejection forces less than 3 kg
  • Ultra low fluid retention
  • Made in USA
Specifications
Volume (Metric) 20 μL
Filtered Non-filtered
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Length (English) 1.9 in.
Length (Metric) 48.3 mm
Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Clean Claims ATP, RNase/DNase, and Endotoxin-free
For Use With (Application) Liquid Handling
Format Refill
Tip Style Flextop
Quantity 1056 Tips/Refill, 10 Refills/Cs.
Shipping Condition RT
ACT Label Yes
Disposable Single-use
For Use With (Equipment) Rainin LTS
Graduated No
Product Line SuperSlik Low Retention
Material Polypropylene Resin