missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUMO1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
302.00€ - 498.00€
Specifications
| Antigen | SUMO1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18274640
|
Novus Biologicals
NBP2-54916 |
100 μL |
498.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18609868
|
Novus Biologicals
NBP2-54916-25ul |
25 μL |
302.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SUMO1 Polyclonal specifically detects SUMO1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| SUMO1 | |
| Polyclonal | |
| Rabbit | |
| Alzheimers Research, Core ESC Like Genes, Neuroscience, Stem Cell Markers | |
| DAP1, GAP modifying protein 1, GAP-modifying protein 1, GMP1SMT3CSMT3H3OFC10UBL1PIC1, SENP2, sentrin, small ubiquitin-related modifier 1, SMT3, SMT3 homolog 3, SMT3 suppressor of mif two 3 homolog 1 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 1 (yeast), Smt3C, SUMO-1, Ubiquitin-homology domain protein PIC1, ubiquitin-like 1 (sentrin), Ubiquitin-like protein SMT3C, Ubiquitin-like protein UBL1 | |
| SUMO1 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 7341 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SDQEAKPSTEDLGDKKEGEYIKLKV | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title