missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUMF2 Monoclonal Antibody (382407)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561308
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
| SUMF2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8BPG6 | |
| SUMF2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Sulfatase Modifying Factor 2/SUMF2, Gln34-Leu308. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 382407 | |
| Unconjugated | |
| SUMF2 | |
| 2610040F05Rik; C-alpha-formyglycine-generating enzyme 2; C-alpha-formylglycine-generating enzyme 2; DKFZp566I1024; DKFZp686I1024; DKFZp686L17160; DKFZp781L1035; FGE2; Inactive C-alpha-formylglycine-generating enzyme 2; MGC99485; Paralog of formylglycine-generating enzyme; paralog of the formylglycine-generating enzyme; pFGE; PSEC0171; sulfatase modifying factor 2; sulfatase-modifying factor 2; Sumf2; UNQ1968/PRO4500 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 67902 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction