missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SULT4A1 Monoclonal Antibody (634630)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561597
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure(number and length of exons) is similar among family members. SULT4A1 isoforms a and b are selectively expressed in brain tissue. Isoform a is the predominant of the two isoforms of this protein. Isoform b includes an alternate segment, compared to isoform a, that causes a frameshift. The resulting protein (isoform b) has a distinct and shorter C-terminus when compared to isoform a.
Specifications
| SULT4A1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9BR01 | |
| SULT4A1 | |
| E. coli-derived recombinant human Cytosolic Sulfotransferase 4A1/SULT4A1, Ala2-Lys284. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 634630 | |
| Unconjugated | |
| SULT4A1 | |
| 2400007A17Rik; AI853543; brain sulfotransferase 4A1; brain sulfotransferase-like protein; BRSTL1; BR-STL-1; DJ388M5.3; family 4A, member 1; hBR-STL; hBR-STL-1; LOW QUALITY PROTEIN: sulfotransferase 4A1; mBR-STL; nervous system cytosolic sulfotransferase; nervous system sulfotransferase; NST; rBR-STL; ST4A1; sulfotransferase 4A1; sulfotransferase family 4A member 1; sulfotransferase family 4A, member 1; sulfotransferase-related protein; SULT4A1; SULTX3; Unknown (protein for MGC:136828); zgc:136828 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 25830 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction