missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur Standard: S @ 750 μg/g (0.0750 wt%) in #2 Diesel Fuel, VHG™
Click to view available options
Quantity:
100 mL
500 mL
1 L
1 gal
Unit Size:
1 GAL
1 L
100mL
500mL
Description
Sulfur Standard: S @ 750 μg/g (0.0750 wt%) in #2 Diesel Fuel
Specifications
Specifications
| Chemical Name or Material | Sulfur |
| CAS | 7704-34-9 |
| Methods | XRF |
| Analyte or Component Names | Sulfur |
| Shipping Condition | Room Temperature |
| Molecular Formula | S |
| Certifications/Compliance | ISO 17034 |
| InChI Formula | InChI=1S/S |
| SMILES | [S] |
| IUPAC Name | sulfur |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction