missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur & Chlorine Standard: S, Cl @ 50 μg/g in 75 cSt Mineral Oil
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
| Chemical Name or Material | Chlorine∣Sulfur |
| CAS | 7782-50-5, 7704-34-9 |
| Analyte or Component Names | Chlorine∣Sulfur |
| Shipping Condition | Room Temperature |
| Solution Type | Mixture |
| Element | Multi-element |
| Solvent or Matrix | 75 cSt Mineral Oil |
| Industry Type | Industrial QC |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Chlorine 50 μg/g∣Sulfur 50 μg/g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction