missing translation for 'onlineSavingsMsg'
Learn More

Sulfur & Chlorine Standard: S, Cl @ 1000 μg/g in 75 cSt Mineral Oil

Product Code. 30327428
Click to view available options
Quantity:
500 mL
800 mL
Unit Size:
500mL
800mL
This item is not returnable. View return policy

Product Code. 30327428

Brand: VHG Labs VHGSCLOIL1000800

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorine∣Sulfur
CAS 7782-50-5, 7704-34-9
Analyte or Component Names Chlorine∣Sulfur
Shipping Condition Room Temperature
Solution Type Mixture
Element Multi-element
Solvent or Matrix 75 cSt Mineral Oil
Industry Type Industrial QC
Quantity 800 mL
Concentration or Composition (by Analyte or Components) Chlorine 1000 μg/g∣Sulfur 1000 μg/g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.