Learn More
Sulfotyrosine Antibody (7C5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP2-59384
 View more versions of this product
Product Code. 18245833
  • 669.00€ / 100µg
Description
Sulfotyrosine Monoclonal specifically detects Sulfotyrosine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sulfotyrosine
Monoclonal
Unconjugated
PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide
Synthetic Sulfotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH).
100 μg
Primary
Non-species specific
Purified
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
7C5
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50
Mouse
Protein G purified
RUO
Specific for sulfated tyrosine residues on various proteins. Does not detect free Sulfotyrosine. Does not cross-react with phosphotyrosine.
Store at -20C.