missing translation for 'onlineSavingsMsg'
Learn More

Sulfotep 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30328502
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328502 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328502 Supplier Dr. Ehrenstorfer Supplier No. DREA17010000AL1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfotep
CAS 3689-24-5
Analyte or Component Names Sulfotep
Shipping Condition +20°C
Molecular Formula C8 H20 O5 P2 S2
Concentration 1000 μg/mL
Certifications/Compliance ISO/IEC 17025
Synonym Sulfotep, Tetraethyl dithionopyrophosphate
InChI Formula InChI=1S/C8H20O5P2S2/c1-5-9-14(16,10-6-2)13-15(17,11-7-3)12-8-4/h5-8H2,1-4H3
Solution Type Single Solution
SMILES CCOP(=S)(OCC)OP(=S)(OCC)OCC
IUPAC Name diethoxyphosphinothioyloxy-diethoxy-sulfanylidene-λ^{5}-phosphane
Solvent or Matrix Acetonitrile
Industry Type Environmental Testing
Molecular Weight (g/mol) 322.32
Quantity 1 mL
Formula Weight 322.0227 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.