Learn More
Sulfaclozine 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Artikelnummer. 30317924
Ansicht ändern
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30317924 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30317924 Supplier Dr. Ehrenstorfer Supplier No. DREA16990290AL1000
 Restricted Item
Estimated Shipment: 07-01-2027
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sulfachloropyrazine
CAS 102-65-8
Analyte or Component Names Sulfachloropyrazine
Shipping Condition +20°C
Molecular Formula C10 H9 Cl N4 O2 S
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Benzenesulfonamide, 4-amino-N-(6-chloropyrazinyl)- (9CI), Sulfanilamide, N1-(6-chloropyrazinyl)- (7CI,8CI), 4-Amino-N-(6-chloro-2-pyrazinyl)benzenesulfonamide, Esb3, Esb3 (antibiotic), N1-(6-Chloropyrazinyl)sulfanilamide, Sulfachloropyrazine, Sulfaclozine, Sulfatyf
InChI Formula InChI=1S/C10H9ClN4O2S/c11-9-5-13-6-10(14-9)15-18(16,17)8-3-1-7(12)2-4-8/h1-6H,12H2,(H,14,15)
SMILES Nc1ccc(cc1)S(=O)(=O)Nc2cncc(Cl)n2
IUPAC Name 4-amino-N-(6-chloropyrazin-2-yl)benzenesulfonamide
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 284.72
Quantity 1 mL
Formula Weight 284.0135 g/mol