Learn More
Abnova™ SUB1 (Human) Recombinant Protein
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
- Sequence: APEKPVKKQKTGETSRALSSSKQSSSSRDDNMFQIGKMRYVSVRDFKGKVLIDIREYWMDPEGEMKPGRKGISLNPEQWSQLKEQISDIDDAV
Specifications
Specifications
| Accession Number | AAH09610.1 |
| Gene ID (Entrez) | 10923 |
| Name | SUB1 homolog (S. cerevisiae) |
| Preparation Method | Wheat germ expression system |
| Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
| Quantity | 25 μg |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Gene Alias | MGC102747, P15, PC4, p14 |
| Gene Symbol | SUB1 |
| Species | Wheat Germ (in vitro) |
| Show More |