missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STYXL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00€ - 590.10€
Specifications
| Antigen | STYXL1 |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18120518
|
Novus Biologicals
NBP2-47611 |
0.1 mL |
624.00€ 590.10€ / 0.10mL Save 33.90€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639575
|
Novus Biologicals
NBP2-47611-25ul |
25 μL |
280.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STYXL1 Polyclonal specifically detects STYXL1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| STYXL1 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 51657 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PGLLLCEPTELYNILNQATKLSRLTDPNYLCLLDVRSKWEYDESHVITALRVKKKNNEYLLPESVDLECVKYCVVYDNNSSTLEILLKDDDDDSDSDGDGKD | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| dual specificity phosphatase 24 (putative), Dual specificity phosphatase inhibitor MK-STYX, Dual specificity protein phosphatase 24, DUSP24, Map kinase phosphatase-like protein MK-STYX, MKSTYX, MK-STYX, serine/threonine/tyrosine interacting-like 1, serine/threonine/tyrosine-interacting-like protein 1 | |
| STYXL1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title