missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STXBP5 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Specifications
Antigen | STXBP5 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16862985
|
Proteintech
24512-1-AP-20UL |
20 μL |
140.00€
20µL |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
16852985
|
Proteintech
24512-1-AP-150UL |
150 μL |
399.00€
150µL |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
Syntaxin 1 is a component of the 7S and 20S SNARE complexes which are involved in docking and fusion of synaptic vesicles with the presynaptic plasma membrane. This gene encodes a syntaxin 1 binding protein. In rat, a similar protein dissociates syntaxin 1 from the Munc18/n-Sec1/rbSec1 complex to form a 10S complex, an intermediate which can be converted to the 7S SNARE complex. Thus this protein is thought to be involved in neurotransmitter release by stimulating SNARE complex formation. Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
STXBP5 | |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat | |
Q5T5C0, Q9WU70 | |
134957, 81022 | |
STXBP5 Fusion Protein Ag21629 | |
Primary | |
-20°C | |
STXBP5 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
LGL3, LLGL3, Nbla04300, STXBP5, Syntaxin binding protein 5, Tomosyn 1 | |
Stxbp5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |