missing translation for 'onlineSavingsMsg'
Learn More

STXBP5 Rabbit anti-Human, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 24512-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16862985

  • 140.00€ / 20µL
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

Syntaxin 1 is a component of the 7S and 20S SNARE complexes which are involved in docking and fusion of synaptic vesicles with the presynaptic plasma membrane. This gene encodes a syntaxin 1 binding protein. In rat, a similar protein dissociates syntaxin 1 from the Munc18/n-Sec1/rbSec1 complex to form a 10S complex, an intermediate which can be converted to the 7S SNARE complex. Thus this protein is thought to be involved in neurotransmitter release by stimulating SNARE complex formation. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications

Specifications

STXBP5
Polyclonal
Unconjugated
STXBP5
LGL3, LLGL3, Nbla04300, STXBP5, Syntaxin binding protein 5, Tomosyn 1
Rabbit
Antigen Affinity Chromatography
RUO
134957, 81022
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5T5C0, Q9WU70
Stxbp5
STXBP5 Fusion Protein Ag21629
20 μL
Primary
Rat, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers