missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STX17 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
129.00€ - 399.00€
Specifications
| Antigen | STX17 |
|---|---|
| Clone | 4G3 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235216
|
Proteintech
81899-1-RR-100UL |
100 μL |
439.00€ 399.00€ / 100µL Save 40.00€ 9% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236124
|
Proteintech
81899-1-RR-20UL |
20 μL |
149.00€ 129.00€ / 20µL Save 20.00€ 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The STX17 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human STX17. This antibody recognizes human, mouse, rat antigen. The STX17 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| STX17 | |
| 1000 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| 55014 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 4G3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| FLJ20651; MGC102796; MGC126613; MGC126615 | |
| IgG | |
| Protein A purification | |
| Recombinant Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title