Learn More
Thermo Scientific™ Stripping Buffer, 4X
Product Code. 15453609
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
Product Code. 15453609
Brand: Thermo Scientific™ J60925.AK
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Western Blot Stripping Buffer
Form Liquid
Quantity 250 mL
Concentration 4X
Content And Storage Ambient temperatures