missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Striatin Polyclonal antibody specifically detects Striatin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Striatin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | MGC125642, SG2NA, striatin, striatin, calmodulin binding protein, striatin, calmodulin-binding protein |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 581-780 of human STRN (NP_003153.2). HQRLLSCSADGTLRLWNTTEVAPALSVFNDTKELGIPASVDLVSSDPSHMVASFSKGYTSIFNMETQQRILTLESNVDTTANSSCQINRVISHPTLPISITAHEDRHIKFYDNNTGKLIHSMVAHLEAVTSLAVDPNGLYLMSGSHDCSIRLWNLESKTCIQEFTAHRKKFEESIHDVAFHPSKCYIASAGADALAKVFV |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?