missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Striated Muscle Myosin Heavy Chain Antibody (83B6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50254-0.025ml
This item is not returnable.
View return policy
Description
Striated Muscle Myosin Heavy Chain Monoclonal antibody specifically detects Striated Muscle Myosin Heavy Chain in Human, Mouse, Rat, Chicken, Rabbit samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Striated Muscle Myosin Heavy Chain | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Mouse | |
| Ascites | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| 83B6 | |
| Unconjugated | |
| Ascites | |
| Purified human skeletal muscle myosin | |
| 0.025 mL | |
| Cardiovascular Biology, Cellular Signaling | |
| Human, Mouse, Rat, Chicken, Rabbit | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction