missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Protein Name: Streptavidin
- Source: Streptomyces avidinii
- Expression System: Native
- Tested Applications: Flow Cyt, FRET
- Purification: Size-exclusion chromatography
- Form: Lyophilized; includes 0.01% Sodium azide, 0.88% Sodium chloride, 0.121% Tris, 0.1% BSA, pH 8.2
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 0.88% sodium chloride, 0.121% Tris, 0.1% BSA, 0.01% sodium azide, pH8.2 |
| Content And Storage | Store at -20°C. |
| Form | Lyophilized |
| Label or Dye | APC |
| Shipping Condition | Blue Ice |
| Substrate Properties | Protein-based Substrate |
| Target Molecule | Biotin |
| Formulation | Lyophilized, pH 8.2, 0.01% sodium azide, 0.88% sodium chloride, 0.121% Tris, 0.1% BSA |
| Quantity | 1 mg |
| For Use With (Application) | Flow Cytometry, FRET |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?