Learn More
Novus Biologicals™ Streptavidin Check&Go! Control Kit
Product Code. 18048163 Shop All Bio Techne Products
Click to view available options
Quantity:
30 Tests
Unit Size:
30 tests
This product is for research use only and is not approved for use in humans or in clinical diagnosis
Product Code. 18048163
Brand: Novus Biologicals™ 40040030
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This product is for research use only and is not approved for use in humans or in clinical diagnosis
Control kit
  • Components:
    • 30 Protein A/G strips
    • 2 Cryovials of 10x Running Buffer
    • 1 Vial of 40nm InnovaCoat Gold Biotin
    • 1 Vial of Positive Control
  • This product is for research use only and is not approved for use in humans or in clinical diagnosis
  • Kits are guaranteed for 6 months from date of receipt
TRUSTED_SUSTAINABILITY
Specifications
Kit Contents 30 protein A/G strips, 1 vial of 40 nm InnovaCoat Gold biotin, 1 vial of positive control, 2 cryovials of 10X running buffer
Quantity 30 Tests