Learn More
IBA Lifesciences™ Strep-Tactin™ XT BLI Coupling Kit for Coating of Biosensors
Product Code. 17580655 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17580655 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17580655 Supplier IBA Lifesciences™ Supplier No. 24380000
  • ONLINE EXCLUSIVE PRICE
    947.00€
    825.00€ / 1 sets
    Save 122.00€
    12% Off
 Restricted Item
Estimated Shipment: 28-10-2026
The XT BLI Coupling Kit can be used for coating BLI biosensors for reversible capture and analysis of proteins.
Strep-Tactin™ XT BLI Coupling Kit is intended for site-directed, reversible capture of Twin-Strep-tag™ fusion proteins for biomolecular interaction analysis using Bio-Layer Interferometry (BLI). The pM affinity of Strep-Tactin™ XT for Twin-Strep-tag™ fusion proteins allows specific capture even from complex samples, such as culture supernatants. Coated biosensors can be easily regenerated by application of the provided Regeneration Buffer without loss of the Strep-Tactin™ XT coating, which enables multiple uses.
  • Provides Strep-Tactin™ XT, Immobilization Buffer, Ethanolamine, EDC, and s-NHS sufficient for coating of 96 biosensors (1 tray)
Specifications
Binding Strength In Low pM Range
Detection Method Optical Biosensing Technology
For Use With (Equipment) Bio-Layer Interferometry (BLI) Systems
High-throughput Compatibility Yes
Quantity 1 Kit
For Use With (Application) Biomolecular Interaction Analysis
Ligand Type Strep-Tactin™ XT
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • strep-tactinXT BLI coupling kit for interaction analysis
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: tris, EDTA