Learn More
IBA Lifesciences Strep-Tactin™ TACS Agarose Column
Product Code. 17227750 Shop All IBA Lifesciences Products
Change view
Click to view available options
Capacity (Metric):
0.3 mL
1 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17227750 1 mL Each
17287590 0.3 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17227750 Supplier IBA Lifesciences Supplier No. 66310001
  • ONLINE EXCLUSIVE PRICE
    116.00€
    100.00€ / Pack of 1
    Save 16.00€
    13% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Gravity flow column suitable for traceless affinity cell and exosome isolation
  • Strep-Tactin™ TACS Agarose columns contain agarose macrobeads coated with the streptavidin variant Strep-Tactin™.
  • A special crosslinking permits the use for cell isolation and depletion approaches.
  • They were developed for column-based Traceless Affinty Cell Selection (Fab-TACS™/Nano-TACS™).
  • In this approach, Fab fragments fused to a Twin-Strep-tag™ (Fab-Streps) or nanobodies (Nano-Streps) are bound to Strep-Tactin™ on the agarose matrix.
  • After specifically capturing cells, Fab-/Nano-Streps are released from the Strep-Tactin™ Agarose by biotin addition.
  • This way the cells are eluted ready for further downstream applications.
  • The same principle is applicable for exosome isolation (Fab-TACS™ for exosomes).
  • Alternatively to Fab-Streps/Nano-Streps, it is possible to load columns with Twin-Strep-tagged, desthiobiotinylated or biotinylated antibodies (irreversible binding).
  • This makes Strep-Tactin™ TACS Agarose columns also suitable for negative cell selection or depletion approaches.
Specifications
Formulation Pre-packed in buffer, pH 8: 100 mM Tris-HCl pH 8.0, 1 mM EDTA, 150 mM NaCl, 0.01% sodium azide
Support Type Cross-linked agarose
For Use With (Application) Affinity Chromatographic Cell or Exosome Isolation
Capacity (Metric) 1 mL
For Research Use Only.