Learn More
IBA Lifesciences Strep-Tactin™ Superflow™ High Capacity Column
Product Code. p-7230582 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 mL
5 mL
5 x 0.2 mL
Unit Size:
1mL
Pack of 5
Product Code. 12856466
Brand: IBA Lifesciences 21209550
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Gravity flow column to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
  • Strep-Tactin™ Superflow™ high capacity columns contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™ and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
  • In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™ Superflow™ high capacity has a higher density of immobilized Strep-Tactin™, which leads to a 3- to 5-fold higher yield of purified protein.
  • To reduce the waiting period during sample and buffer application, the use of the application aid WET FRED is recommended.
Specifications
Binding Capacity 150 to 500 nmoL
Capacity (Metric) 1 mL
Support Type Superflow 6
Quantity 5 x 0.2 mL
For Use With (Application) One-Step Purification of Recombinant Strep-Tag Proteins
Type Gravity Flow Column
For Research Use Only.