missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRA8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
230.00€ - 317.00€
Specifications
| Antigen | STRA8 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18666812
|
Novus Biologicals
NBP2-98692-100ul |
100 μg |
317.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18632052
|
Novus Biologicals
NBP2-98692-50ul |
50 μg |
230.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STRA8 Polyclonal antibody specifically detects STRA8 in Human samples. It is validated for Western BlotSpecifications
| STRA8 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| stimulated by retinoic acid gene 8 homolog (mouse), stimulated by retinoic acid gene 8 protein homolog | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human STRA8. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 346673 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title