missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STING/TMEM173 Antibody (2922D) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-18817
This item is not returnable.
View return policy
Description
STING/TMEM173 Monoclonal antibody specifically detects STING/TMEM173 in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| STING/TMEM173 | |
| Monoclonal | |
| 1.0 mg/ml | |
| Western Blot 1 to 2 μg/mL, Simple Western, Immunohistochemistry 0.5 μg/mL, Immunocytochemistry/Immunofluorescence 3 μg/mL | |
| anti-STING, endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, hSTING, human STING, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, mouse STING, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, SAVI, Stimulator of interferon genes protein, stimulator of interferon protein, sting, STING monoclonal, STING-beta, TMEM173, transmembrane protein 173 | |
| Partial recombinant protein made to amino acids 215-379 of human TMEM173/STING (UniProt Q86WV6). | |
| 0.1 mg | |
| Cancer, Immunology, Innate Immunity | |
| 340061 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 2922D | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction