Learn More
CEEC Controlled Environment Equipment Corporation™ Sticky Mats
Eliminate dust contamination from soles with CEEC Controlled Environment Equipment Corp.™ Sticky Mats. They adhere directly to a floor or in an aluminum frame for decontaminating shoes upon entering a cleanroom environment.
85.00€ - 275.00€
Product Code | Brand | Color | Dimensions (L x W) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Dimensions (L x W) | Price | Quantity & Availability | ||||
11836383
|
CEEC Controlled Environment Equipment Corporation™
177636 |
Blue | 45 x 114 cm |
85.00€
Pack of 120 |
In Stock
Log in to see stock. |
||||
11876373
|
CEEC Controlled Environment Equipment Corporation™
177352 |
White | 45 x 91 cm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
11886373
|
CEEC Controlled Environment Equipment Corporation™
177423 |
White | 45 x 114 cm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
11816383
|
CEEC Controlled Environment Equipment Corporation™
CSS183630FBB |
Blue | 46 x 91cm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
11826383
|
CEEC Controlled Environment Equipment Corporation™
177405 |
Blue | 66 x 114 cm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
11866373
|
CEEC Controlled Environment Equipment Corporation™
CSS243634TWW |
White | 61 x 91 cm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
11846383
|
CEEC Controlled Environment Equipment Corporation™
177528 |
Blue | 66 x 91 cm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
Description
- Adhesive dust mats
- Sheets numbered from 30 to 1
- To stick directly to the floor or in an aluminum frame