Learn More
CEEC Controlled Environment Equipment Corporation™ Sticky Mats
Product Code. 11836383
Click to view available options
Color:
Blue
White
Dimensions (L x W):
45 x 91 cm
46 x 91cm
61 x 91 cm
66 x 91 cm
45 x 114 cm
66 x 114 cm
Unit Size:
Pack of 120
Product Code. 11836383
Brand: CEEC Controlled Environment Equipment Corporation™ 177636
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Eliminate dust contamination from soles with CEEC Controlled Environment Equipment Corp.™ Sticky Mats. They adhere directly to a floor or in an aluminum frame for decontaminating shoes upon entering a cleanroom environment.
  • Adhesive dust mats
  • Sheets numbered from 30 to 1
  • To stick directly to the floor or in an aluminum frame
Specifications
Dimensions (L x W) 45 x 114 cm
Color Blue