Learn More
CEEC Controlled Environment Equipment Corporation™ Sticky Mats
Eliminate dust contamination from soles with CEEC Controlled Environment Equipment Corp.™ Sticky Mats. They adhere directly to a floor or in an aluminum frame for decontaminating shoes upon entering a cleanroom environment.
Brand: CEEC Controlled Environment Equipment Corporation™ 177528
Additional Details : Weight : 3.60000kg
Description
- Adhesive dust mats
- Sheets numbered from 30 to 1
- To stick directly to the floor or in an aluminum frame
Specifications
66 x 91 cm | |
Blue | |
120/Pk. |