Specifications
| Content And Storage | Please see the StepOne™ to StepOnePlus™ Upgrade User Bulletin (sku 4401093) for upgrade instructions. This kit contains necessary components to support the upgrade from a 48-well StepOne™ Real Time PCR System to a 96-well system. |
| For Use With (Equipment) | StepOne™ |
| Product Line | StepOnePlus |
| Quantity | 1 Upgrade |