Learn More
Cytiva Steel Spanner Wrenches
Product Code. p-8007006 Shop All Cytiva Products
Click to view available options
For Use With (Equipment):
Locking Nuts 95 to 100 mm dia.
Nuts 58 to 62 mm dia.
Unit Size:
Each
Product Code. 12465670
Brand: Cytiva 28964778
Only null left
Specifications
Type Spanner Wrench
Material Steel with galvanized surface
Capacity (Metric) 95 to 100 μL
Quantity 1/Pk.
For Use With (Equipment) Locking Nuts 95 to 100 mm dia.