missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEAP3/TSAP6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95115-0.02ml
This item is not returnable.
View return policy
Description
STEAP3/TSAP6 Polyclonal antibody specifically detects STEAP3/TSAP6 in Mouse samples. It is validated for Western Blot
Specifications
| STEAP3/TSAP6 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| dudlin 2, dudlin-2, dudulin 2, dudulin-2, Dudulin-2,1010001D01Rik, EC 1.16.1.-, hpHyde, hTSAP6, metalloreductase STEAP3, pHyde, six transmembrane prostate protein 3, Six-transmembrane epithelial antigen of prostate 3, STEAP family member 3, TSAP6STMP3, tumor suppressor pHyde, Tumor suppressor-activated pathway protein 6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human STEAP3 (NP_060704.2). MPEEMDKPLISLHLVDSDSSLAKVPDEAPKVGILGSGDFARSLATRLVGSGFKVVVGSRNPKRTARLFPSAAQVTFQEEAVSSPEVIFVAVFREHYSSLCSLSDQLAGKILVDVSNPTEQEHLQHRESNAEYLASLFPTCTVVKAFNVISAWTLQAGPRDGNRQVPICGDQPEAKRAVSE | |
| 0.02 mL | |
| Apoptosis, Cancer, Cell Biology, Endocrinology, Signal Transduction | |
| 55240 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction