missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEAP1 Antibody (1057323) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5587M
This item is not returnable.
View return policy
Description
STEAP1 Monoclonal antibody specifically detects STEAP1 in Human samples. It is validated for Flow Cytometry,Western Blot
Specifications
| STEAP1 | |
| Monoclonal | |
| DyLight 594 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Western Blot | |
| 1057323 | |
| Western Blot, Flow Cytometry | |
| EC 1.16.1.-, PRSS24MGC19484, six transmembrane epithelial antigen of the prostate, six transmembrane epithelial antigen of the prostate 1, Six-transmembrane epithelial antigen of prostate 1, STEAPmetalloreductase STEAP1 | |
| E. coli-derived recombinant human STEAP-1, Met1-Trp71, Accession # Q9UHE8 | |
| 0.1 mL | |
| Cancer | |
| 26872 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction