Learn More
STCH Antibody (2F6), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-60243MFV500
Product Code. 18477175
  • 562.00€ / 0.10mL
Description
STCH Monoclonal antibody specifically detects STCH in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ELISA, Immunocytochemistry
Mouse
6782
IgG1 κ
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Recombinant human STCH (23-471aa) purified from E. coli