missing translation for 'onlineSavingsMsg'
Learn More

STAT5a/b [p Tyr699, p Tyr694] Antibody (1247C), CoraFluor™ 1, Novus Biologicals™

Product Code. 30145703 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30145703 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30145703 Supplier Novus Biologicals Supplier No. FAB41901CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

STAT5a/b Monoclonal antibody specifically detects STAT5a/b in Human samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAT5a/b
Applications Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 1247C
Conjugate CoraFluor 1
Formulation PBS
Gene Alias MGF, STAT5, STAT5A signal transducer and activator of transcription 5A
Host Species Rabbit
Immunogen Phosphopeptide containing human STAT5b Y699 site (amino acid sequence of this peptide is identical to a corresponding region of human STAT5a containing Y694), Accession # P51692
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6776
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.