missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
STAT3 Monoclonal antibody specifically detects STAT3 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry,KnockDown
Specifications
Specifications
| Antigen | STAT3 |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry, KnockDown |
| Classification | Monoclonal |
| Clone | CL0490 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulation | Lyophilized in 5% Trehalose. |
| Gene Alias | Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor) |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?