missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT2 Antibody (S01-2F6), Novus Biologicals™
Rabbit Monoclonal Antibody
444.00€ - 680.00€
Specifications
| Antigen | STAT2 |
|---|---|
| Clone | S01-2F6 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot, ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18621834
|
Novus Biologicals
NBP3-20074-100ul |
100 μg |
680.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671555
|
Novus Biologicals
NBP3-20074-50ul |
50 μg |
444.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STAT2 Monoclonal antibody specifically detects STAT2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Sandwich ELISASpecifications
| STAT2 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| interferon alpha induced transcriptional activator, ISGF-3, MGC59816, p113, signal transducer and activator of transcription 2, signal transducer and activator of transcription 2, 113kD, signal transducer and activator of transcription 2, 113kDa, STAT113 | |
| A synthetic peptide of mouse STAT2 (Uniprot # Q9WVL2) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S01-2F6 | |
| Western Blot, ELISA, Sandwich ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 6773 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title