missing translation for 'onlineSavingsMsg'
Learn More

STAT1, p Ser727 Antibody (PSM1), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody

531.00€

Specifications

Antigen STAT1
Clone PSM1
Applications Western Blot
Classification Monoclonal
Conjugate Janelia Fluor 549
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18437414
View Documents
Novus Biologicals
NB500-515JF549
0.1 mL
531.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

STAT1 Monoclonal antibody specifically detects STAT1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications

Specifications

STAT1
Western Blot
Janelia Fluor 549
Mouse
Apoptosis, Breast Cancer, Cancer, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
50mM Sodium Borate
6772
IgG1
Protein A purified
PSM1
Monoclonal
Purified
RUO
Human
DKFZp686B04100, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1
STAT1 peptide sequence 721-733 (Ser727 phosphorylated).
Primary
Store at 4°C in the dark.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.