Learn More
IBA Lifesciences StarGate pPSG-IBA Acceptor Vectors for E. coli Expression
Product Code. 15603768 Shop All IBA Lifesciences Products
Click to view available options
Vector:
pPSG-IBA 105
pPSG-IBA 123
pPSG-IBA 164
pPSG-IBA 168
pPSG-IBA 23
pPSG-IBA 25
pPSG-IBA 3
pPSG-IBA 33
pPSG-IBA 5
pPSG-IBA 62
pPSG-IBA 63
pPSG-IBA 64
pPSG-IBA 65
pPSG-IBA wt1
pPSG-IBA103
pPSG-IBA143
pPSG-IBA145
pPSG-IBA162
pPSG-IBA167
pPSG-IBA35
pPSG-IBA43
pPSG-IBA45
Unit Size:
5µg
Product Code. 15603768
Brand: IBA Lifesciences 54243001
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Achieves high-level expression in E. coli by the T7 promoter by providing specified genetic surroundings for optimal expression and purification of non-toxic proteins.
pPSG-IGA E. coli expression vectors:
pPSG-IBA vectors use the T7 promoter and T7 RNA polymerase for high-level transcription of the gene of interest.
  • As the T7 promoter is stronger than the tet promoter, pPSG-IBA vectors are recommended in cases where expression with the tet promoter does not lead to significant yields of the recombinant protein.
    • High-level expression in E. coli by bacteriophage T7 promoter
    • High-level transcription by T7 RNA polymerase in BL21 strains
    • High-level expression of non-toxic proteins
    • Induction by IPTG
    • Suitable for in vitro transcription/translation
    StarGate System
    IBA developed StarGate to provide a simple, fast and highly efficient cloning process for genes of interest (GOI) for use in functional expression, purification, detection, immobilization or separation of recombinant proteins:
    • Minimal extra modification of the gene due to short combinatorial sites
    • Inherent high-level cloning efficiency due to a directed reaction (no equilibrium)
    • GOIs are cloned to obtain a Donor Vector; 2 or more genes can be combined using Fusion Cloning
    • Genes are then transferred into Acceptor Vectors that provide optimal genetic surroundings
    • The final expression vector, the Destination Vector, is then placed into the respective host
  • Specifications
    Format Liquid
    For Use With (Equipment) StarGate™ System, E. Coli BL21(DE3)
    Inducing Agent E. coli host with T7 RNA polymerase gene (e.g. BL21(DE3))
    pH 8
    Promoter T7
    Product Type Acceptor Vector for E. coli expression
    Storage Buffer 20 μL TE buffer: 10 mm Tris-HCl, 1 mm EDTA / 5 μg
    Content And Storage 2°C to 8°C for frequent usage, -20°C for long-term storage
    Protein Tag Strep-tag for C-terminal and His Tag (6x) for N-terminal
    Concentration 250 ng/μL
    Quantity 5 μg
    Vector pPSG-IBA43
    Color Clear
    For Use With (Application) Cloning genes of interest for use in functional expression, purification, detection, immobilization or separation of recombinant proteins