missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ STARD3 Recombinant Protein

Human STARD3 full-length ORF recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00010948-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16105912

  • 528.00€ / 25µg

Estimated Shipment: 17-07-2023
to see stock.
Add to basket

Description

Description

This gene encodes a member of a subfamily of lipid trafficking proteins that are characterized by a C-terminal steroidogenic acute regulatory domain and an N-terminal metastatic lymph node 64 domain. The encoded protein localizes to the membranes of late endosomes and may be involved in exporting cholesterol. Alternate splicing results in multiple transcript variants.

  • Theoretical MW (kDa): 74.69
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Sequence: MSKLPRELTRDLERSLPAVASLGSSLSHSQSLSSHLLPPPEKRRAISDVRRTFCLFVTFDLLFISLLWIIELNTNTGIRKNLEQEIIQYNFKTSFFDIFVLAFFRFSGLLLGYAVLQLRHWWVIAVTTLVSSAFLIVKVILSELLSKGAFGYLLPIVSFVLAWLETWFLDFKVLPQEAEEERWYLAAQVAVARGPLLFSGALSEGQFYSPPESFAGSDNESDEEVAGKKSFSAQEREYIRQGKEATAVVDQILAQEENWKFEKNNEYGDTVYTIEVPFHGKTFILKTFLPCPAELVYQEVILQPERMVLWNKTVTACQILQRVEDNTLISYDVSAGAAGGVVSPRDFVNVRRIERRRDRYLSSGIATSHSAKPPTHKYVRGENGPGGFIVLKSASNPRVCTFVWILNTDLKGRLPRYLIHQSLAATMFEFAFHLRQRISELGARA

Best when used within three months from the date of receipt.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH08747
Solution
10948
STARD3 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
STARD3
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
74.69
8
Glutathione Sepharose 4 Fast Flow
25 ug
MSKLPRELTRDLERSLPAVASLGSSLSHSQSLSSHLLPPPEKRRAISDVRRTFCLFVTFDLLFISLLWIIELNTNTGIRKNLEQEIIQYNFKTSFFDIFVLAFFRFSGLLLGYAVLQLRHWWVIAVTTLVSSAFLIVKVILSELLSKGAFGYLLPIVSFVLAWLETWFLDFKVLPQEAEEERWYLAAQVAVARGPLLFSGALSEGQFYSPPESFAGSDNESDEEVAGKKSFSAQEREYIRQGKEATAVVDQILAQEENWKFEKNNEYGDTVYTIEVPFHGKTFILKTFLPCPAELVYQEVILQPERMVLWNKTVTACQILQRVEDNTLISYDVSAGAAGGVVSPRDFVNVRRIERRRDRYLSSGIATSHSAKPPTHKYVRGENGPGGFIVLKSASNPRVCTFVWILNTDLKGRLPRYLIHQSLAATMFEFAFHLRQRISELGARA
CAB1/FLJ41370/MLN64/es64
STARD3
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers