missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STARD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
187.00€
Specifications
| Antigen | STARD3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
STARD3 Polyclonal specifically detects STARD3 in Rat samples. It is validated for Western Blot.Specifications
| STARD3 | |
| Western Blot 1 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Lipid and Metabolism | |
| PBS and 1 mg/ml BSA with 0.05% Sodium Azide | |
| 10948 | |
| Recombinant MLN64 protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Rat | |
| CAB1, es64, Metastatic lymph node gene 64 protein, metastatic lymph node protein 64, MLN 64, MLN64FLJ41370, Protein CAB1, StARD3, StAR-related lipid transfer (START) domain containing 3, stAR-related lipid transfer protein 3, START domain containing 3, START domain-containing protein 3, steroidogenic acute regulatory protein related | |
| STARD3 | |
| IgG | |
| Affinity Purified | |
| MLN 64 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title