Learn More
Starch Indicator Solution, Chem Lab™
Product Code. 11957443
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
500mL
Each
Product Code. 11957443
Brand: Chem-Lab CL07.2601.0500
Specifications
Melting Point 0°C
Solubility Information Soluble in water
Odor Odorless
Color Colorless
pH ±7
Quantity 500 mL
Physical Form Liquid