missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Staphylococcus Aureus Enterotoxin B Antibody (S643), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
519.00€
Specifications
| Antigen | Staphylococcus Aureus Enterotoxin B |
|---|---|
| Clone | S643 |
| Applications | Sandwich ELISA |
| Classification | Monoclonal |
| Conjugate | Janelia Fluor 646 |
Description
Staphylococcus Aureus Enterotoxin B Monoclonal antibody specifically detects Staphylococcus Aureus Enterotoxin B in Bacteria samples. It is validated for ELISASpecifications
| Staphylococcus Aureus Enterotoxin B | |
| Sandwich ELISA | |
| Janelia Fluor 646 | |
| Mouse | |
| Bacteria | |
| S. aureus | |
| IgG1 | |
| Protein G purified |
| S643 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Sodium Borate | |
| Hybridoma clone has been derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with recombinant S. aureus enterotoxin B expressed in E. coli. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title