Learn More
Stanozolol, Lipomed
Product Code. 30459841
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459841 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30459841 Supplier Lipomed Supplier No. LPMSTA152250
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Stanozolol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Stanozolol
CAS 10418-03-8
Analyte or Component Names Stanozolol
Shipping Condition Room Temperature
Molecular Formula C21 H32 N2 O
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES C[C@]1(O)CC[C@H]2 [C@@H]3 CC[C@H]4 Cc5[nH]ncc5C[C@]4(C)[C@H]3 CC[C@]12 C
IUPAC Name (1 S,2 S,10 S,13 R,14 S,17 S,18 S)-2,17,18-trimethyl-6,7-diazapentacyclo[11.7.0.0 ^{2,10}0.0 ^{4,8}0.0 ^{14,18}]icosa-4(8),5-dien-17-ol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 328.49
Quantity 50 mg
Formula Weight 328.2515 g/mol