missing translation for 'onlineSavingsMsg'
Learn More

Standard 1508: Cl @ 75μg/g; Zn, Mo @ 750μg/g in 75 cSt Oil

Product Code. 30326923
Click to view available options
Quantity:
400 g
Unit Size:
400g
This item is not returnable. View return policy

Product Code. 30326923

Brand: VHG Labs VHGJET1508400G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorine∣Molybdenum∣Zinc
CAS 7782-50-5, 7439-98-7, 7440-66-6
Analyte or Component Names Chlorine∣Molybdenum∣Zinc
Shipping Condition Room Temperature
Solution Type Mixture
Element Multi-element
Solvent or Matrix 75 cSt Mineral Oil
Industry Type Industrial QC
Quantity 400 g
Concentration or Composition (by Analyte or Components) Chlorine 75 μg/g∣Molybdenum 750 μg/g∣Zinc 750 μg/g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.