Learn More
Gilson™ Stainless Steel Frits
Product Code. 12975680
Change view
Click to view available options
Porosity:
2 μm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Porosity Unit Size
12975680 2 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12975680 Supplier Gilson™ Supplier No. 0201079
  • ONLINE EXCLUSIVE PRICE
    19.00€
    15.18€ / Pack of 1
    Save 3.82€
    20% Off
Estimated Shipment: 28-01-2027
Specifications
Diameter (Metric) Outer 0.96 mm
Diameter (Metric) Ring 3.17 mm
Material Stainless Steel
Porosity 2 μm
Thickness (Metric) 0.76 mm
Type Frit