Learn More
Gilson™ Stainless Steel Frit
Product Code. 12923200
Change view
Click to view available options
Porosity:
20 μm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Porosity Unit Size
12923200 20 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12923200 Supplier Gilson™ Supplier No. 4271159
  • ONLINE EXCLUSIVE PRICE
    38.00€
    33.42€ / Pack of 1
    Save 4.58€
    12% Off
Estimated Shipment: 22-12-2026
Specifications
Diameter (Metric) Frit 1.9 cm
Diameter (Metric) Ring 2.24 cm
Material Stainless Steel
Porosity 20 μm
Thickness (Metric) 0.16 cm
Type Frit